Learn More
Nanocs BSA PEG Conjugate, 5mg
Supplier:  Nanocs PG5KBS
Encompass
BSA PEG conjugate was made from by the covalent attachment of PEG 2000 to the BSA molecule via its lysine residue. Conjugate is provided in lyophilized powder form.
Catalog No. NC1587188