Learn More
Medchemexpress LLC BSB (trans,trans)-1-bromo-2,5-bis-(3-hydroxycarbonyl-4-hydroxy)styrylbenzene | 291766-06-8 | 97.2% | 481.29 g/mol | C24H17BrO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD144310MG
Encompass_Preferred
BSB is a Congo red-derived fluorescent probe that selectively binds extracellular amyloid-β and intracellular aggregates of tau and synuclein; it is used as a prototype imaging agent in Alzheimer's disease research. Supplied as a purified small-molecule solid with supporting documentation for research use.
  • Binds extracellular amyloid-β and intracellular tau and synuclein aggregates.
  • High purity suitable for biochemical and imaging applications.
  • Provided as a small-molecule solid with recommended dissolution in DMSO (use newly opened DMSO; warming/sonication may aid solubility).
  • Store at 4°C protected from light; in solution store at -80°C for long-term stability.
  • Comes with datasheet, certificate of analysis, and safety data sheet for research use.
Catalog No. 50-002-97145