Learn More
Research Products International Corp AEBSF, 10 MG
SDP
Supplier:  Research Products International Corp A200100.01
Encompass_Preferred
Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.
Catalog No. 50-197-8643