Learn More
Apexbio Technology LLC AEBSF.HCl, 100mg. Cas: 30827-99-7 MFCD: MFCD00132962. Serine protease inhibitor
SDP
Supplier:  Apexbio Technology LLC A2573100
Encompass_Preferred
AEBSF is a broad spectrum, irreversible inhibitor of serine proteases [1].AEBSF is a covalently binding inhibitor of proteases, including trypsin, chymotrypsin, plasmin and thrombin. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4746