Learn More
Medchemexpress LLC BSJ-02-162 (CDK4/6-IN-11) | 2139329-47-6 | 98.1% | C43H49N11O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14499510MG
Encompass_Preferred
BSJ-02-162 (CDK4/6-IN-11) is a proteolysis-targeting chimera (PROTAC) developed for preclinical research to induce ubiquitin-mediated degradation of cyclin-dependent kinases 4 and 6. The molecule combines an E3 ligase-recruiting thalidomide-derived moiety with a palbociclib-derived target ligand connected by a modular linker to promote selective proteasomal degradation of CDK4/6.
  • Induces targeted degradation of CDK4 and CDK6 via PROTAC mechanism.
  • High chemical purity suitable for research applications.
  • Available as a solid powder with recommended cold storage.
  • Soluble in DMSO at high concentration; ultrasonic assistance may be required.
  • Supplied in multiple small pack sizes for screening and optimization.
Catalog No. 50-002-60382