Learn More
Medchemexpress LLC BSJ-03-204 | 2349356-09-6 | 99.8% | 832.90 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1362501MG
Encompass_Preferred
BSJ-03-204 is a PROTAC connected by ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader (PROTAC), with IC50s of 26.9 nM for CDK4/D1 and 10.4 nM for CDK6/D1. BSJ-03-204 does not induce IKZF1/3 degradation and exhibits anti-cancer activity.
  • Potent anti-proliferative effects on MCL cell lines.
  • Potently induces a G1 arrest.
  • Selectively degrades CDK4/6 in WT cells, not IKZF1/3.
Catalog No. 50-003-51883