Learn More
Medchemexpress LLC BSJ-03-204 | 2349356-09-6 | 99.8% | 832.90 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY136250
Encompass_Preferred
BSJ-03-204 is a PROTAC that connects ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader (PROTAC), with IC50s of 26.9 nM for CDK4/D1 and 10.4 nM for CDK6/D1. This compound does not induce IKZF1/3 degradation and exhibits anti-cancer activity.
  • Potent and selective CDK4/6 dual degrader
  • Does not cause IKZF1/3 degradation
  • Demonstrates anti-cancer activity
  • Potent anti-proliferative effects on MCL cell lines
  • Induces G1 arrest
Catalog No. 50-003-47851