Learn More
Medchemexpress LLC BSJ-03-204 (triTFA) | 2349356-09-6 | 99.8% | 832.90 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13625050MG
Encompass_Preferred
BSJ-03-204 is a PROTAC connected by ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader with anti-cancer activity, and does not induce IKZF1/3 degradation.
  • Potent anti-proliferative effects on MCL cell lines
  • Potently induces a G1 arrest
  • Selective degradation of CDK4/6 in WT cells
Catalog No. 50-003-51161