Learn More
Medchemexpress LLC BSJ-04-132 | 2349356-39-2 | 99.1% | C42H49N11O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13625210MG
Encompass_Preferred
BSJ-04-132 is a Ribociclib-based PROTAC designed to induce selective degradation of cyclin-dependent kinase 4 (CDK4) through cereblon recruitment. It is supplied as a high-purity research reagent for biochemical and cell-based studies of CDK4 regulation and targeted protein degradation mechanisms.
  • Selective CDK4 degrader suitable for mechanistic studies.
  • Designed to recruit cereblon for ubiquitin-proteasome degradation.
  • High stated purity for reproducible experimental results.
  • Available in small-mg packaging for research use.
  • Compatible with biochemical and cell-based assays.
Catalog No. 50-002-61646