Learn More
Medchemexpress LLC HY-136252 5mg Medchemexpress, BSJ-04-132 CAS:2349356-39-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1362525MG
Encompass_Preferred
Medchemexpress, HY-136252 5mg BSJ-04-132 CAS:2349356-39-2 BSJ-04-132 is a potent and selective Ribociclib-based CDK4 degrader (PROTAC), with IC50s of 50.6 nM and 30 nM for CDK4/D1 and CDK6/D1, respectively. BSJ-04-132 does not induce CDK6 and IKZF1/3 degradation. BSJ-04-132 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1428