Learn More
New England Biolabs, Inc. BsmFI – 500 units
SDP
Supplier:  New England Biolabs, Inc. R0572L
Encompass
A restriction endonuclease that recognizes the sequence GGGACNNNNNNNNNN^NNNN_.
Catalog No. 50-812-267