Learn More
Bioworld BSSS, 50 mg
Supplier:  Bioworld 218100011
Encompass_Preferred
Bis(sulfosuccinimidyl)suberateA popular non-cleavable, membrane-impermeable, water-soluble DSS analogCategory: Amine-reactiveReactive Groups: sulfo-NHS-esterReactive Toward: amino gropsWater Soluble: YesMembrane Permeable: NoCleavability: Non-cleavableMW: 572.43
Catalog No. 50-199-249