Learn More
ATCC BT; Turbinate; Bovine (Bos taurus)
Supplier:  ATCC CRL1390
Encompass
BT is a cell that was isolated from the turbinate of a cow. This cell line has been tested and found negative for the Bovine viral diarrhea virus (BVDV). This cell line was deposited by AW McClurkin.
Catalog No. 50-238-3474