Learn More
Apexbio Technology LLC BT2 34576-94-8 10mg
SDP
Supplier:  Apexbio Technology LLC BA332910
Encompass_Preferred
BT2 (CAS 34576-94-8) is a small-molecule inhibitor of branched-chain -ketoacid dehydrogenase kinase (BCKDK) By binding to BCKDK BT2 induces conformational changes in the N-terminal domain promoting dissociation from the branched-chain -ketoacid dehydrogenase complex (BCKDC) BT2 suppresses BCKDK activity with an IC50 of 3 19 M and demonstrates selective inhibition as indicated by an IC50 of 59 M in relevant assays This compound serves as a valuable tool for investigating BCKDK-regulated metabolic pathways and the role of BCKDK in branched-chain amino acid metabolism
Catalog No. 50-000-88782