Learn More
Medchemexpress LLC BT2 500mg | 34576-94-8 | 500 MG

Supplier: Medchemexpress LLC HY114855500MG
3,6-dichlorobenzo[b]thiophene-2-carboxylic acid (BT2, CAS 34576-94-8) is a research reagent used as an allosteric inhibitor of branched-chain α-ketoacid dehydrogenase kinase (BDK) to modulate branched-chain amino acid metabolism in biochemical studies. It is supplied as a solid powder for laboratory research.
- Allosteric BDK inhibitor for biochemical research.
- High purity, typically >98% (HPLC).
- Chemical formula C9H4Cl2O2S, molecular weight 247.10 g/mol.
- Supplied as a solid powder; store at 0-4 °C short-term or -20 °C long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.