Learn More
Medchemexpress LLC HY-114855 50mg Medchemexpress, BT2 CAS:34576-94-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11485550MG
Encompass_Preferred
Medchemexpress, HY-114855 50mg BT2 CAS:34576-94-8 BT2 is a BCKDC kinase (BDK) inhibitor with an IC50 of 3.19 μM. BT2 binding to BDK triggers helix movements in the N-terminal domain, resulting in the dissociation of BDK from the branched-chain α-ketoacid dehydrogenase complex (BCKDC). BT2 (compound 4) is also a potent and selective Mcl-1 inhibitor with a Ki value of 59 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9375