Learn More
ATCC BT-20; Breast Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB19
Encompass
BT-20 cells were isolated in 1958 from a breast tumor of a 74-year-old, White, female breast cancer patient. Use these cells in your breast cancer and immuno-oncology research.
Catalog No. 50-238-2385