Learn More
Medchemexpress LLC BT44 RET Activator | 924759-42-2 | C28H27F4N3O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1531755MG
Encompass_Preferred
BT44 is a selective RET activator that can penetrate the blood-brain barrier. It is suitable for research into neurodegenerative disorders and diabetes mellitus.
  • Promotes RET phosphorylation and selectively activates downstream cascades
  • Promotes neurite outgrowth from sensory neurons
  • Alleviates sensory signs in neuropathic pain models
  • Protects IB4-positive neurons
  • Reverses amphetamine-induced motor imbalance in Parkinson's disease models
  • Penetrates the blood-brain barrier
Catalog No. 50-003-73157