Learn More
Strem, An Ascensus Company CAS 186598-40-3. 1g. Bis(t-butylamino)silane 97% BTBAS. MFCD03411245
SDP
Supplier:  Strem, An Ascensus Company 1410601G
Encompass
CAS 186598-40-3. 1g. Bis(t-butylamino)silane 97% BTBAS. MFCD03411245. Molecular Weight 174.36. Molecular Formula [NH(C4H9)]2SiH2. Color/form colorless liq. Strem 14-1060. http//www.strem.com/catalog/v/14-1060/
Catalog No. 50-211-0714