Learn More
TARGETMOL CHEMICALS INC BTdCPU 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T71501MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. BTdCPU is a potent activator of heme-regulated eIF2alpha kinase (HRI) .BTdCPU promotes eIF2alpha phosphorylation and induced apoptosis in resistant cell. Purity 98.61%
Catalog No. 50-287-7856