Learn More
Medchemexpress LLC BTK Protein, Human (Active, sf9, His) | 94% | Approximately 78 kDa | 20 UG
Supplier: Medchemexpress LLC HYP7204820UG
This BTK Protein, Human (Active, sf9, His) is a recombinant human protein produced in Sf9 insect cells, featuring an N-10*His tag. It is supplied as a lyophilized powder with high purity and confirmed biological activity, suitable for research applications.
- Approximately 78 kDa molecular weight
- 94% purity as determined by reducing SDS-PAGE
- Lyophilized from a 0.22 μm filtered solution
- Biologically active with specific activity >35 nmol/min/mg
- Suitable for various research applications