Learn More
Chemglass Life Sciences Bottle, washing, self-venting, LDPE, 500 mL - wide mouth, leak-proof closure
SDP
Supplier:  Chemglass Life Sciences CG1131V002
Encompass_Preferred
This 500 mL laboratory wash bottle is made from low-density polyethylene (LDPE) with a molded, leak-resistant closure and stem. It includes a non-metallic self-vent to reduce solvent leakage, a wide mouth for easier filling, and an angled tip that can be trimmed to adjust flow.
  • 500 mL capacity.
  • Low-density polyethylene body.
  • Polypropylene closure and molded stem for leak resistance.
  • Non-metallic self-vent reduces solvent leakage.
  • Wide mouth for easy filling.
  • Angled neck and trim-to-adjust tip for controlled dispensing.
  • Pack quantity: pack of 6.
Catalog No. NC2959220