Learn More
BTNX Inc BTNX CLIA-Waived Drug Test Cup 12-Panel with Adulteration Strip (Flat Cup Format) (12.26) AMP1000 BUP10 BZO300 COC300 EDDP100 ETG500 FYL10 MDMA500 MET1000 MOP300 OXY100 THC50 + Cre/SG/pH
Supplier:  BTNX Inc D12.261VAF
Encompass_Preferred
The Rapid Response 12 Parameter Multi-Drug One Step Cup (Flat) + Adulteration Test detects AMP1000 BUP10 BZO300 COC300 EDDP100 ETG500 FYL10 MDMA500 MET1000 MOP300 OXY100 THC50 + Cre/SG/pH in urine samples. It has a longer detection window than saliva tests and provides results in 5 minutes. Quantity: 25/kit. CLIA-Waived.Rapid Response 12 Parameter Multi-Drug One Step Cup (Flat) + Adulteration Test is compatible with our RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork.
Catalog No. NC3957449