Learn More
BTNX Inc BTNX GHB Test Card 2/Kit Gamma-Hydroxybutyric Acid (GHB) Cutoff 500µg/mL
Supplier:  BTNX Inc GHB8CD1582
Encompass_Preferred
The Rapid Response Spiked Drink Test Card detects GHB at 500 µg/mL in beverages. Quantity: 2/kit. Time to result: 60 seconds.
Catalog No. NC3957518