Learn More
BTNX Inc BTNX Single Drug Test Strip 100/Kit Marijuana (THC) Cutoff 50 ng/mL
Supplier:  BTNX Inc THC1S13100
Encompass_Preferred
The Rapid Response Single Drug Test Strip for Marijuana (Cutoff 50 ng/mL) is a rapid, qualitative test for the detection of a selected drug in human urine. Our Single Drug Test Strips are straightforward, targeted, and economic drug screening solution. Quantity: 100/kit. Time to Result: 5 minutes.
Catalog No. NC3957413