Learn More
Medchemexpress LLC BTTES | 1334179-86-0 | C19H32N10O4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15341950MG
Encompass_Preferred
BTTES is a tris(triazolylmethyl)amine-based ligand for Cu(I) without apparent toxicity. It improves the cycloaddition reaction rapidly in living systems.
  • Tris(triazolylmethyl)amine-based ligand for Cu(I)
  • No apparent toxicity
  • Improves cycloaddition reaction rapidly in living systems
Catalog No. 50-003-73375