Learn More
Selleck Chemical LLC BTZ043 Racemate 10mg 957217-65-1
SDP
Supplier:  Selleck Chemical LLC S109710MG
Encompass_Preferred
BTZ043 racemate is a decaprenylphosphoryl-?-D-ribose 2'-epimerase (DprE1) inhibitor acting as a new antimycobacterial agent that kill Mycobacterium tuberculosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-827-5