Learn More
Medchemexpress LLC BTZO-1 | 99420-15-2 | 99.3% | 240.28 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY11008410MG
Encompass_Preferred
BTZO-1 is a chemical probe designed to bind to Macrophage migration inhibitory factor (MIF), with a Kd value of 68.6 nM, specifically requiring the N-terminal Pro1 for its binding. This compound demonstrates the ability to activate antioxidant response element (ARE)-mediated gene expression and effectively suppress oxidative stress-induced cardiomyocyte apoptosis in vitro.
  • Suppresses serum deprivation-induced cardiomyocyte death at concentrations as low as 16 nM.
  • Upregulates mRNA protein levels of GST Ya and HO-1 in cardiomyocytes.
  • Suitable for research into neurological, eye or ear, inflammation, blood, cardio-cerebrovascular, digestive system, and neurodegenerative diseases.
  • Available in solid form and solutions in DMSO.
  • Stable for 3 years at -20°C as powder.
Catalog No. 50-003-44245