Learn More
Cayman Chemical Buca javnca L M. Bucn D 10mg
SDP
Supplier:  Cayman Chemical 4014010 MG
Encompass_Preferred
A quassinoid with diverse biological activities; active against chloroquine-resistant P. falciparum (IC50 = 0.015 UG/ml); selectively inhibits the proliferation of PANC-1, SW 1990, and Capan-1 cancer cells over Hs68 fibroblasts (IC50s = 2.53, 5.21, 1.35, and >30 µM, respectively); protects against MPTP-induced motor impairments and dopaminergic neurodegeneration in a mouse model of Parkinson’s disease at 20 or 40 mg/kg; decreases carrageenan-induced paw inflammation in rats at 0.25 or 1 mg/kg
Catalog No. 50-294-2279