Learn More
Boca Scientific Inc Buccal Prep Plus DNA Isolation Kit, 50 reactions, with 50 SK-2S Buccal Swabs
Supplier:  Boca Scientific Inc BPP50/SK2
Encompass
This kit includes the BPP-50 Buccal Prep Plus Isolation Kit and 50 SK-2S buccal swabs for buccal DNA collection and isolation. SK-2S swabs are specially designed for maximum DNA yields due to a unique swab matrix and quick release surface. Each SK-2S buccal swab is packaged with a 2ml plastic collection tube and special cap. The Buccal-Prep Plus DNA Isolation Kit is a precipitation-based kit combining all the necessary optimized buffers to fully lyse and simply purify DNA from buccal swabs. The kit has a fast protocol that produces high yields and purity for all downstream processing applications. No solvents, alcohol, or other external buffers are required, giving fewer processing steps and creating a faster process time.
Catalog No. NC1894505