Learn More
Chemglass Life Sciences Buchner Funnel, Table Top, Polyethylene, Perforated, 24in ID, Removable Plate
SDP
Supplier:  Chemglass Life Sciences CG141322
Encompass_Preferred
Buchner Funnel, Table Top, Polyethylene, Perforated, 24in ID, Removable Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Removable hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Removable plate is for use in precious metals recovery; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).
Catalog No. CG141322