Learn More
Chemglass Life Sciences Buchner Funnel, Table Top, Polyethylene, Porous, 10.25in ID, Medium Porosity, Fixed Plate
SDP
Supplier:  Chemglass Life Sciences CG141301
Encompass_Preferred
Buchner Funnel, Table Top, Polyethylene, Porous, 10.25in ID, Medium Porosity, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Welded hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel. Filter plates are supported by a multiple ring grid; HDPE, 6.4mm (1/4") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used with porous filter plates. Medium potosity is 45-90 microns; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).
Catalog No. CG141301