Learn More
Medchemexpress LLC HY-B0764 50mg , Bucladesine (sodium) CAS:16980-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB076450MG
Encompass_Preferred
HY-B0764 50mg Bucladesine (sodium) CAS: 16980-89-5 Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1145