Learn More
Medchemexpress LLC Buclizine dihydrochloride | 129-74-8 | 99.6% | C28H35Cl3N2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0128A100MG
Encompass_Preferred
Buclizine dihydrochloride is an orally active antihistamine antiallergic compound known for its anti-tumor activity. It is used in research to study cellular processes.
  • Orally active antihistamine antiallergic compound
  • Exhibits anti-tumor activity
  • Inhibits growth of MCF-7 cells
  • Arrests cell cycle in G1 phase in a dose-dependent manner
  • Decreases expression of TCTP and cell cycle regulatory proteins in MCF-7 cells
  • Increases expression of pro-apoptotic MCL-1S
Catalog No. 50-004-32414