Learn More
Apexbio Technology LLC Buclizine HCl 129-74-8 50mg
SDP
Supplier:  Apexbio Technology LLC B154650
Encompass_Preferred
Buclizine hydrochloride (CAS 129-74-8) is a synthetic piperazine derivative classified as an antihistamine and anticholinergic agent It functions by competitively inhibiting histamine H1 receptors thereby reducing histamine-mediated responses and also exhibits antagonistic activity at muscarinic acetylcholine receptors These dual actions have established its use in studies exploring the modulation of allergic reactions motion sickness and the central and peripheral effects of histaminergic and cholinergic pathways Buclizine HCl is utilized in biomedical research to investigate receptor pharmacology and signal transduction mechanisms associated with histaminergic and cholinergic neurotransmission
Catalog No. 50-000-89925