Learn More
Medchemexpress LLC Buddlejasaponin IVb | 152580-79-5 | 99.8% | 943.12 | C48H78O18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN213810MG
Encompass_Preferred
Buddlejasaponin IVb is a triterpene saponin isolated from Clinopodium chinense that exhibits hemostatic activity, shortening thrombin time (TT) by approximately 20.6%. The compound is supplied as a purified solid with molecular formula C48H78O18 and molecular weight 943.12 g/mol. Typical purity is 99.8% (HPLC). It is intended for biochemical and pharmacological research applications and is provided in small research-scale quantities; follow recommended storage conditions to preserve activity.
  • High purity suitable for analytical and in vitro studies.
  • Documented hemostatic activity for coagulation research.
  • Available in small research-scale quantities for dose-response studies.
  • Provided as a dry solid and as a DMSO solution format for immediate use.
  • Stable under recommended storage conditions to preserve activity.
Catalog No. 50-002-95610