Learn More
Medchemexpress LLC Budesonide (22R)-enantiomer | 51372-29-3 | MFCD00870255 | 98.0% | 430.53 g/mol | C25H34O6 | 1MG

Supplier: Medchemexpress LLC HY1259221MG
(22R)-Budesonide is the (22R)-enantiomer of budesonide, a non-halogenated glucocorticoid with high local anti-inflammatory activity. It is supplied for research use to study glucocorticoid receptor interactions and stereochemistry-dependent pharmacology.
- Chemical formula: C25H34O6.
- Molecular weight: 430.53 g/mol.
- CAS number: 51372-29-3.
- Typical reported purity: 98.0%.
- Intended for research use only.
- Consult the certificate of analysis for storage and detailed purity data.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.