Learn More
Research Products International Corp Budget-Solve Complete Counting Cocktail, 4 Liters
SDP
Supplier:  Research Products International Corp 111167
Encompass_Preferred
Budget-Solve is an economical ready-to-use cocktail for liquid scintillation counting. No dilution is required. It contains a blend of quench resistant fluors in an efficient emulsifier system. Budget-Solve is an economical product of exceptional performance. It is designed to handle a wide range of liquid scintillation counting samples.
WARNING! This product can expose you to Methyl alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-983
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov