Learn More
Enzo Life Sciences Bufalin (10mg). CAS: 465-21-4
SDP
Supplier:  Enzo Life Sciences BMLAP3030010
Encompass_Preferred
Cardiotonic steroid specific Na+/K+-ATPase inhibitor. More potent than ouabain (BML-CM109). Induces cell differentiation. Induces apoptosis in U937 and HL-60 cells by activation of AP-1 via a MAPK pathway that involves JNK. Regulates Tiam1 gene. Down regulates expression of 14-3-3 protein. Alternative name: (3beta,5beta)-3,14-Dihydroxybufa-20,22-dienolide, 5beta,20(22)-Bufadienolide-3beta,14-diol. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-0313