Learn More
Abcam Bufalin, Na+/K+-ATPase inhibitor, 10MG
Supplier:  Abcam AB14563610MG
Encompass_Preferred
MW 386.5 Da, Purity >98%. Na+/K+-ATPase inhibitor. Potent steroid receptor coactivators SRC-3 and SRC-1 inhibitor. Induces cell differentiation and apoptosis. Activates AP-1 via JNK. Shows antitumor effects *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5091