Learn More
Selleck Chemical LLC Bufexamac S3023-50mg
SDP
Supplier:  Selleck Chemical LLC S302350MG
Encompass_Preferred
Bufexamac(Bufexamic acid) is a COX inhibitor for IFN- release with EC50 of 8 9 M
Catalog No. 50-311-1575