Learn More
Ethos Biosciences Hematology Buffer, pH 6.8, 1gal, Ethos
Supplier:  Ethos Biosciences 5548G
Encompass_Preferred
Hematology Buffer pH 6.8 is a buffered solution designed for hematology staining procedures. Commonly used with Romanowsky-type stains, provides the optimal pH environment for proper dye interaction and accurate cellular differentiation. The solution helps ensure consistent staining quality, clear nuclear and cytoplasmic detail, and reliable microscopic evaluation. Ideal for peripheral blood smears and bone marrow preparations, it supports routine diagnostic and research workflows.
Catalog No. 50-005-74354