Learn More
As One International Inc Buffer kit, 10x Ammonium Buffer, 10x Standar Buffer, 10x Combination Buffer all detergent anf Magnesium free and 25 mM MgCl2, 1 x 1.5 ml each
Supplier:  As One International Inc AO306501
Encompass_Preferred
Ammonium Buffer is recommended for most PCR applications. It results in high yield of PCR products and minimizes the need for optimization of magnesium concentration or primer annealing temperatures. Ammonium Buffer also works well with difficult templates like GC rich DNA sequences.Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required.Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures. Magnesium free buffers are recommended if you want to optimize the magnesium concentration.Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry.
Catalog No. 50-165-5983