Learn More
Cole-Parmer Oakton Traceable pH Standard Buffer with Calibration, Clear, pH 7.416; 500 mL
Supplier:  Cole-Parmer 0065546
Encompass_Preferred
Choose from clear or color-coded standards from common calibration points. NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory. Oakton pH buffers are compared against, and are traceable to, NIST Standard Reference Materials. Each bottle includes a NIST-traceable calibration report stating accuracy at 25C to 0.010 pH. Store at room temperature. Certified Reference Materials (CRM) meet federal, state, and or local agencies strictest mandates, and deliver exact calibration results for any pH meter.
Catalog No. 50-196-1732