Learn More
Hach Company Buffer Powder Pillows, pH 10.01 Color-coded Blue, pk/50
Supplier:  Hach Company 2227166
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Color-coded for easy identification (Blue). pH is 10.01 ±0.02 at 25°C. Pack of 50 Powder Pillows
Catalog No. 50-199-5578