Learn More
Hach Company Buffer Powder Pillows, pH 7.00 Color-coded Yellow, pk/250
Supplier:  Hach Company 2227064
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Color-coded for easy identification (Yellow). pH is 7.00 ±0.02 at 25°C. Pack of 250 Powder Pillows
Catalog No. 50-199-5574