Learn More
Hach Company Buffer Powder Pillows, pH 9.00, pk/50
Supplier:  Hach Company 1410766
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder Pillow prepares 50 mL of fresh buffer solution. Colorless. pH is 9.00 ±0.02 at 25°C. Pack of 50 Powder Pillows. Pack of 50 Powder Pillows
Catalog No. 50-199-4929