Learn More
Hach Company Buffer Powder Pillows pH 4.01
Supplier:  Hach Company 2226964
Encompass
Buffer Powder Pillows-pH 4 01 Color-coded Red-pk/250pH Buffer Powder Pillows for pH electrode calibration Each powder pillow prepares 50 mL of fresh buffer solution Color-coded for easy identification (Red) pH is 4 01 0 02 at 25 C Pack of 250 Powder Pillows
Catalog No. 50-002-07018