Learn More
Hach Company Buffer Solution, pH 7.00, 4 L
Supplier:  Hach Company 1222256
Encompass
pH Buffer Standard Solution for pH electrode calibration. Colorless pH buffer solution. pH is 7.00 ±0.02 at 25°C. 4 L bottle
Catalog No. 50-199-4861