Learn More
Hach Company Buffer Solution, Phosphate, for Chlorine, 500 mL
Supplier:  Hach Company 2335549
Encompass
For chlorine determination by the DPD ferrous titration method
Catalog No. 50-199-5826